Z663107
Kern EW series Toploader balances
Kern EW 6000-1M + 963-128, DKD Calibration Certificate included
Synonym(s):
laboratory balance, weighing balance
About This Item
Recommended Products
AC/DC input
230 V
material
stainless steel weighing plate
feature
CE compliant
DKD Calibration Certificate included
packaging
pack of 1 ea
manufacturer/tradename
Kern EW 6000-1M + 963-128
availability
not available in USA and Canada
weighing capacity measuring range
6,000 g, resolution: 0.1 g, linearity: ±0.2 g
W × depth × H
180 mm × 230 mm × 85 mm
platform size
172 mm × 142 mm
weight
2 kg
battery power
rechargeable
Looking for similar products? Visit Product Comparison Guide
General description
- Large LCD display
- Capacity display
- Protective working cover
- Test weight
- 3 years warranty
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service