SAB4700896
Monoclonal Anti-CD3 zeta (PhosphoTyr111) antibody produced in mouse
clone EM-55
Synonym(s):
CD247, CD3 zeta chain, IMD25, T3Z, TCRz
About This Item
Recommended Products
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
EM-55, monoclonal
form
buffered aqueous solution
species reactivity
human, mouse
concentration
1 mg/mL
technique(s)
flow cytometry: 2 μg/mL
accession no.
P20963
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
phosphorylation (pTyr111)
Gene Information
human ... CD247(919)
General description
Immunogen
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service