Skip to Content
Merck
All Photos(1)

Documents

31153

Sigma-Aldrich

Ammonium metavanadate

puriss. p.a., reag. Ph. Eur., ≥99.5%

Synonym(s):

Ammonium vanadate(V)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4VO3
CAS Number:
Molecular Weight:
116.98
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38180409
PubChem Substance ID:

Agency

USP/NF
reag. Ph. Eur.

grade

puriss. p.a.

Assay

≥99.5%

form

solid

density

2.32 g/cm3 at 25 °C (lit.)

anion traces

chloride (Cl-): ≤20 mg/kg
phosphate (PO43-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Cd: ≤10 mg/kg
Co: ≤20 mg/kg
Cu: ≤10 mg/kg
Fe: ≤10 mg/kg
Ni: ≤20 mg/kg
Pb: ≤20 mg/kg
Zn: ≤10 mg/kg

SMILES string

N.O[V](=O)=O

InChI

1S/H3N.H2O.2O.V/h1H3;1H2;;;/q;;;;+1/p-1

InChI key

YBVKNHXQSRDWAA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium metavanadate is an ammonium salt. It participates as an oxidizing agent for condensation reaction of o-phenylenediamine with substituted aryl aldehydes.

Application

Ammonium metavanadate may be employed in the following studies:
  • As catalyst for the synthesis of various 2-substituted aryl benzimidazoles.
  • As oxidizing agent in the determination of chlorpromazine, promethazine, trimeprazine and perphenazine in pharmaceutical samples by flow injection method.
  • Synthesis of dioxidovanadium(V) complex.
  • Preparation of Vanadia modified titania (V2O5/TiO2) photo-catalysts.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diptipriya Sethi et al.
Journal of photochemistry and photobiology. B, Biology, 144, 68-74 (2015-03-03)
Vanadia modified titania (V₂O₅/TiO₂) photo-catalysts are prepared by incipient wet impregnation method using aqueous ammonium metavanadate and anatase (Aldrich) titania. Titania with various loading concentrations of vanadia from 0 to 10 wt.% have been prepared and characterized by X-ray diffraction
Simona Nica et al.
Journal of inorganic biochemistry, 147, 193-203 (2015-03-10)
The Schiff-base ligand (H2salhyhNH3)Cl (1) derived from salicylaldehyde and 6-aminohexanoic acid hydrazide hydrochloride reacts with ammonium metavanadate in methanol solution to yield the dioxidovanadium(V) complex [VO2(salhyhNH3)] (2). The utilized hydrazone ligand contains a flexible and protonated amino side chain. Crystallization
Flow injection method for the assay of phenothiazine neuroleptics in pharmaceutical preparations using ammonium metavanadate.
Sultan SM.
Analyst, 116(2), 177-181 (1991)
C Faría de Rodríguez et al.
Investigacion clinica, 39 Suppl 1, 87-97 (1998-07-03)
The present research show, an statistical study of the effects of ammonium metavanadate (AMV) on size, weight and number of offsprings of 56 albino swiss mice (male and female) was carried out. These mice ingested AMV in concentration of 100
Mansour S Abdul Galil et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 67(1), 76-82 (2006-08-22)
A rapid, simple, and most economical spectrophotometric method was proposed for the determination of nitrite in various water samples, soil samples, and roots of leguminous plants. The method is based on decolorizing effect of nitrite on complex formed between hydrogen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service