Skip to Content
Merck
All Photos(1)

Key Documents

90267

Sigma-Aldrich

Lithium citrate tribasic solution

BioUltra, 1.5 M in H2O

Synonym(s):

Citric acid trilithium salt solution, Trilithium citrate solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5Li3O7
CAS Number:
Molecular Weight:
209.92
MDL number:
UNSPSC Code:
12161700

product line

BioUltra

concentration

1.5 M in H2O

impurities

insoluble matter, passes filter test

pH

8.0-9.5 (25 °C, 1.5 M in H2O)

density

1.12 g/mL at 20 °C

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

1.5 M in H2O

UV absorption

λ: 260 nm Amax: 0.040
λ: 280 nm Amax: 0.016

SMILES string

[Li+].[Li+].[Li+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service