Skip to Content
Merck
All Photos(1)

Key Documents

MAB80118

Sigma-Aldrich

Anti-Newcastle Disease Virus Antibody, clone HN14f

clone HN14F, Chemicon®, from mouse

Synonym(s):

NDV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

HN14F, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

isotype

IgG

shipped in

wet ice

General description

Newcastle Disease Virus (NDV) is a member of the paramyxovirus group of enveloped negative-stranded RNA viruses that also includes measles, mumps, respiratory syncytial virus and the parainfluenza viruses.

Specificity

Hemagglutinin-neuraminidase protein of Newcastle virus

Immunogen

Intact, UV-inactivated, egg-grown Newcastle Disease Virus strain Australia-Victoria.

Application

Anti-Newcastle Disease Virus Antibody, clone HN14f detects level of Newcastle Disease Virus & has been published & validated for use in WB.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Format: Purified
Liquid in 0.02M PB with 0.25M NaCl, pH 7.6. Contains 0.1% sodium azide.

Storage and Stability

Store at 2° to 8°C for up to 12 months from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monoclonal antibodies to newcastle disease virus: delineation of four epitopes on the HN glycoprotein
Iorio, R M and Bratt, M A
Journal of virology, 48, 440-450 (1983)
Xiangpeng Ren et al.
Proteome science, 10(1), 32-32 (2012-05-11)
Newcastle disease virus (NDV) is an enveloped RNA virus, bearing severe economic losses to the poultry industry worldwide. Previous virion proteomic studies have shown that enveloped viruses carry multiple host cellular proteins both internally and externally during their life cycle.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service