Skip to Content
Merck
All Photos(1)

Key Documents

HTS192RTA

Sigma-Aldrich

Ready-to-Assay S1P 4 Lysophospholipid Receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.76
Product HTS192RTA is not currently sold in your country. Contact Technical Service

Product Name

Ready-to-Assay S1P 4 Lysophospholipid Receptor Frozen Cells,

biological source

human

Quality Level

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable

NCBI accession no.

detection method

fluorometric

General description

Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

Sphingosine 1-phosphate (S1P) is a biologically active lysophospholipid that transmits signals through a family of five G-protein-coupled receptors to regulate cell proliferation, migration, cytoskeletal organization, and differentiation (Spiegel and Milstien , 2003). S1P4 receptor was reported to couple to multiple subsets of heterotrimeric G proteins (including Gi/o, and G12/13, and possibly Gs). Unlike other S1P receptors, S1P4 expression is restricted in human and mouse to lymph node, spleen, lung, and thymus (Ishii et al. 2001). This expression pattern suggests potential roles of S1P4 in the immune system. Millipore′s cloned human S1P4-expressing cell line is made in the Chem-5 host, which supports high levels of recombinant S1P4 expression on the cell surface and contains high levels of the promiscuous G protein to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists and antagonists at S1P4.

Cell Line Description

  • GPCR Cell Lines
  • Host cells: Chem-5

Application

Human S1P4 / EDG6 GPCR frozen cells for Calcium Flux FLIPR Assays.

Biochem/physiol Actions

  • GPCR Class: A
  • Protein Target: S1P4 / EDG6
  • Target Sub-Family: Lysophospholipid

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial.
Fifty (50) mL of Media Component.

Disclaimer


This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service