Skip to Content
Merck
All Photos(1)

Key Documents

1.09012

Supelco

Nessler′s reagent B

for determination of nitrogen solution B: Sodium hydroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352305

Quality Level

form

liquid

specific analyte(s)

nitrogen

technique(s)

photometry: suitable

pH

14 (20 °C in H2O)

density

1.30 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Application test: conforms

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service