ATE959441737
AstaTech, Inc.
CIS-3-[2-(TERT-BUTOXY)-2-OXOETHYL]-4-METHYLPYRROLIDINE-3-CARBOXYLIC ACID
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
(3S,4R)-3-(2-(tert-butoxy)-2-oxoethyl)-4-methylpyrrolidine-3-carboxylic acid
Inchi Code
1S/C12H21NO4/c1-8-6-13-7-12(8,10(15)16)5-9(14)17-11(2,3)4/h8,13H,5-7H2,1-4H3,(H,15,16)/t8-,12+/m0/s1
InChI key
AYESCXPKJQDVGF-QPUJVOFHSA-N
Purity
95%
Country of Origin
CN
Physical Form
White to Yellow Solid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=112331
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=P17326
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service