AMBH93E4C1B4
Ambeed, Inc.
Spiro[adamantane-2,9'-fluorene]
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
(1r,3r,5r,7r)-spiro[adamantane-2,9'-fluorene]
Inchi Code
1S/C22H22/c1-3-7-20-18(5-1)19-6-2-4-8-21(19)22(20)16-10-14-9-15(12-16)13-17(22)11-14/h1-8,14-17H,9-13H2
InChI key
LGYQERJJBDQQHH-UHFFFAOYSA-N
storage temp.
Sealed in dry,Room Temperature
Purity
98%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
Solid or liquid
Product Link
https://www.ambeed.com/products/727730-31-6.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.ambeed.com/products/727730-31-6.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service