Skip to Content
Merck
All Photos(1)

Documents

720844

Sigma-Aldrich

4-Piperidinopiperidine-1-carbonyl chloride

Synonym(s):

1,4′-Bipiperidinyl-1′-carbonyl chloride, 1-Chlorocarbonyl-4-piperidinopiperidine, [1,4′-Bipiperidine]-1′-carbonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19ClN2O
CAS Number:
Molecular Weight:
230.73
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

mp

60-64 °C

SMILES string

ClC(=O)N1CCC(CC1)N2CCCCC2

InChI

1S/C11H19ClN2O/c12-11(15)14-8-4-10(5-9-14)13-6-2-1-3-7-13/h10H,1-9H2

InChI key

YDNSNQRKIINKPV-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Macrocyclic ureas as selective Chk1 inhibitors
  • Etoposide prodrug for dual prodrug-enzyme antitumor therapy
  • Phosphodiesterase 4 inhibitors
  • Camptothecin analogs

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service