Skip to Content
Merck
All Photos(1)

Key Documents

247073

Sigma-Aldrich

2-Acetyl-5-chlorothiophene

99%

Synonym(s):

5-Chloro-2-thienyl methyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5ClOS
CAS Number:
Molecular Weight:
160.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

117-118 °C/17 mmHg (lit.)

mp

46-49 °C (lit.)

functional group

chloro
ketone

SMILES string

CC(=O)c1ccc(Cl)s1

InChI

1S/C6H5ClOS/c1-4(8)5-2-3-6(7)9-5/h2-3H,1H3

InChI key

HTZGPEHWQCRXGZ-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C S Chidan Kumar et al.
Molecules (Basel, Switzerland), 18(10), 11996-12011 (2013-10-01)
A series of six novel heterocyclic chalcone analogues 4(a-f) has been synthesized by condensing 2-acetyl-5-chlorothiophene with benzaldehyde derivatives in methanol at room temperature using a catalytic amount of sodium hydroxide. The newly synthesized compounds are characterized by IR, mass spectra
Betül Şen et al.
Materials science & engineering. C, Materials for biological applications, 98, 550-559 (2019-03-01)
The reactions of cobalt(II), nickel(II), zinc(II) chlorides and [Pd(DMSO)2Cl2] with 2-acetyl-5-chloro-thiophene thiosemicarbazone (HL) leads to the formation of a series of new complexes: [CoCl2(S-HL)2], 1; [Ni(N,S-L)2], 2 [ZnCl2(S-HL)2], 3 and [PdCl2(N,S-HL)], 4. All the complexes have been characterized by elemental
Chandraju Sadolalu Chidan Kumar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 127, 67-73 (2014-03-19)
The structure of 2-acetyl-5-chlorothiophene (ACT) has been characterized by FT-IR, Raman and single-crystal X-ray diffraction techniques. The isomers, optimized geometric parameters, normal mode frequencies and corresponding vibrational assignments of ACT (C6H5ClOS) have been examined by the density functional theory, with
Ismail Warad et al.
Molecules (Basel, Switzerland), 25(9) (2020-05-15)
Three new tetradentate NNNS Schiff bases (L1-L3) derived from 2-(piperidin-4-yl)ethanamine were prepared in high yields. UV-Visible and FTIR spectroscopy were used to monitor the dehydration reaction between 2-(piperidin-4-yl)ethanamine and the corresponding aldehydes. Structures of the derived Schiff bases were deduced
Khyati P Thakor et al.
Luminescence : the journal of biological and chemical luminescence, 34(1), 113-124 (2019-01-04)
Novel palladium(II) complexes (7a-7e) of substituted quinoline derivatives were synthesized. The complexes were characterized using various techniques such as thermogravimetric analysis (TGA), elemental analysis, conductance measurement, mass, absorption, infra-red (IR), 1 H NMR, 13 C NMR and energy-dispersive X-ray spectroscopy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service