Merck
HomeSmall Molecule HPLCHPLC Discovery HS C18
HPLC Discovery HS C18
Discovery HS C18 HPLC Columns, 120 Å;
3 µm, 5 µm and 10 µm particle sizes available
  • Specifically developed for pharmaceutical analysis and purification
  • Ideal for LC/MS applications - No detectable bleed (as independently tested)
  • Highly stable to ensure excellent run-to-run and lot-to-lot reproducibility and long column life
  • Scalable from analytical to prep
NO DETECTABLE LC/MS BLEED
Discovery HS C18
No TIC Baseline rise.
Essentially no m/z peaks generated from column ("x" indicates peaks also found in blank run)
Column: Discovery HS C18 and Conventional C18, 5cm x 4.6mm, 3µm;
0.1% TFA; Flow Rate: 1mL/min; Temp: ambient, esi+ ion.
Competitor C18
TIC Baseline rise can interfere with sensitivity, and quantitation measurements. Bleed m/z peaks could coincidently interfere with "real" peaks.
Mobile Phase: 15 min. gradient from 0-100% MeOH in
(work conducted by Dr. Shane Needham, Alturas Analytics, Moscow, ID)
(work conducted by Dr. Shane Needham, Alturas Analytics, Moscow, ID)
LOT TO LOT REPRODUCIBILITY OF DISCOVERY HS C18
Column: Discovery HS C18, 15 cm x 4.6 mm, 5 µm
Mobile Phase: 80% 10 mM ammonium acetate, pH 6.8 : 20% MeOH
Flow Rate: 1 mL/min
Temp: 35 °C
Detection: 254 nm;
Injection: 1 µL
1. Uracil (8 µg/mL)
2. Procainamide (30 µg/mL)
3. Sorbic acid (4 µg/mL)
4. Pyridine (100 µg/mL)
5. Caffeine (40 µg/mL)
6. Phenol (240 µg/mL)
g001597_9
GREATER HYDROPHOBIC RETENTION COMPARED TO OTHER POPULAR C18 PHASES
Columns: 7.5 cm x 4.6 mm, 3 µm/3.5 µm
Mobile Phase: 35:65 - H2O:ACN
Flow Rate: 2 mL/min; Detection: 254 nm
g001500
Ordering Information:
3µm Discovery HS C18 HPLC Columns
Sorry, an unexpected error has occurred
Network error: Failed to fetch
5µm Discovery HS C18 HPLC Columns
Sorry, an unexpected error has occurred
Network error: Failed to fetch
10µm Discovery HS C18 HPLC Columns
Sorry, an unexpected error has occurred
Network error: Failed to fetch
2cm Supelguard Cartridges with Discovery HS C18 Packings
Sorry, an unexpected error has occurred
Network error: Failed to fetch
* 569275-U (Kit includes cartridge, stand-alone holder, tubing, 2 nuts and ferrules)