Merck
LC/MS/MS Analysis of Vitamins B1 and B6 on Ascentis® Express OH5
LC/MS/MS Analysis of Vitamins B1 and B6 on Ascentis® Express OH5 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express OH5, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
standard
Product No.
Description
Pricing
Thiamine Hydrochloride (B1)
analytical standard
Pyridoxine Hydrochloride (B6)
analytical standard
CONDITIONS
column
Ascentis Express  OH5, 10 cm x 2.1 mm I.D., 2.7 μm particles (53757-U)
mobile phase
[A] water with 0.1% formic acid/10 mM ammonium formate; [B] acetonitrile with 0.1% formic acid/10 mM ammonium formate; (A:B, 15:85)
flow rate
0.2 mL/min
pressure
1150 psi (79 bar)
column temp.
30 °C
detector
MS, ESI(+), MRM mode
injection
1 μL
sample
10 ng/mL in mobile phase
Description
Analysis Note
The present work establishes an HILIC LC/MS/MS method for analysis of water soluble vitamins B1 and B6 on an Ascentis® Express OH5 column. LC/MS grade solvents were used to supply low background interference and low particulate contaminants for robust, trouble-free operation.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany