Merck
HPLC Analysis of Serotonin Metabolites on Ascentis® C18
HPLC Analysis of Serotonin Metabolites on Ascentis® C18 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® C18 HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
standard
Product No.
Description
Pricing
Tryptophan
Pharmaceutical Secondary Standard; Certified Reference Material
5-Hydroxy-L-tryptophan
phyproof® Reference Substance
5-Hydroxy-L-tryptophan
Pharmaceutical Secondary Standard; Certified Reference Material
CONDITIONS
column
Ascentis C18, 15 cm x 4.6 mm I.D., 5 μm particles (581324-U)
mobile phase
[A] 10 mM formic acid/acetate (pH 3.0): [B] methanol; (80:20, A:B)
flow rate
0.7 mL/min
column temp.
35 °C
detector
ESI (+); overlay of extracted ion chromatograms of individual (M+H)+ species
injection
10 μL
sample
25 mg/L in 90:10, 10 mM formic acid/acetate (pH 3.0):methanol
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany