GC Analysis of a 19-Component PCB Congener Mix on SLB®-IL100
![GC Analysis of a 19-Component PCB Congener Mix on SLB®-IL100 suitable for GC](/deepweb/assets/sigmaaldrich/product/images/561/379/aa2cb74d-5577-4cdd-958d-aa52136222b8/800/aa2cb74d-5577-4cdd-958d-aa52136222b8.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-IL100, 30 m x 0.25 mm I.D., 0.20 μm (28884-U)
oven
60 °C (1 min), 8 °C/min to 230 °C
inj. temp.
250 °C
MSD interface
220 °C
scan range
m/z = 95-500
carrier gas
helium, 1.5 mL/min constant
injection
1 μL splitless (1.0 min)
liner
4 mm I.D., single taper
sample
PCB congener mix, each analyte at 2.5 ppm in n-hexane
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany