GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
sample/matrix
50 ppb each component in 5mL of water
purge trap
VOCARB 3000
purge
40 mL/min for 11 min at ambient temperature
desorption temp.
210 °C for 2 min
bake
260 °C for 10 min
transfer line/valve temp.
110 °C
column
SPB-624, 60 m x 0.32 mm ID, 1.8 μm (24251)
oven
35 °C (8 min), 5 °C/min to 100 °C, 15 °C/min to 220 °C (5 min)
inj. temp.
230 °C
scan range
m/z 41-400
carrier gas
helium, 1.2 mL/min constant
injection
split ~18:1
instrument
OI 4560
Description
Legal Information
SPB is a registered trademark of Merck KGaA, Darmstadt, Germany