Merck
GC Analysis of Polyunsaturated Fatty Acid (PUFA) Methyl Esters on PAG
GC Analysis of Polyunsaturated Fatty Acid (PUFA) Methyl Esters on PAG suitable for GC
Materials
analytical column
Product No.
Description
Pricing
PAG Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
PAG, 30 m × 0.25 mm I.D., 0.25 μm (24223)
oven
220 °C
inj. temp.
250 °C
detector
FID, 260 °C
carrier gas
helium, 25 cm/sec @ 220 °C
injection
1 μL, split 100:1
sample
Omegawax Test Mix (48476)