GC Analysis of Industrial Solvents on Equity-1701
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Equity-1701, 30 m x 0.32 mm I.D., 1.0 μm (28199-U)
oven
35 °C (8 min), 4 °C/min to 130 °C (2 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 25 cm/sec
injection
0.5 μL 200:1 split
liner
Split type, cup design
sample
59-Component neat solvent mix
Description
Legal Information
null