Merck
GC Analysis of a 32-Component Anilines Mix on SLB®-IL59
GC Analysis of a 32-Component Anilines Mix on SLB®-IL59 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-IL59 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.20 μm
CONDITIONS
column
SLB-IL59, 30 m x 0.25 mm I.D., 0.20 μm (28891-U)
oven
75 °C (2 min), 10 °C/min to 250 °C, 15 °C/min to 300 °C (25 min)
inj. temp.
250 °C
detector
MSD, interface 275 °C, m/z = 30-300
carrier gas
helium, 0.7 mL/min constant
injection
1 μL pulsed splitless (25 psi pulse)
liner
4 mm I.D., split/splitless type, single taper design
sample
Each analyte at 50 μg/mL in methylene chloride
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany