Merck
  • Frequency of Pro475Ser polymorphism of ADAMTS13 gene and its association with ADAMTS-13 activity in the Korean population.
Frequency of Pro475Ser polymorphism of ADAMTS13 gene and its association with ADAMTS-13 activity in the Korean population.
Yonsei medical journal (2008-06-27)
Mun Ju Jang, Nam Keun Kim, So Young Chong, Hye Jin Kim, Seon Ju Lee, Myung Seo Kang, Doyeun Oh
ABSTRACT
The in vitro study suggested that proline to serine polymorphism in codon 475 (C1423T) of the A Disintegrin and Metalloprotease with ThromboSpondin type 1 repeats-13 (ADAMTS-13) gene is related to reduced activity of ADAMTS-13. In this study, the frequency of the Pro475Ser polymorphism in Koreans was studied and plasma ADAMTS-13 activity was measured to find out whether this polymorphism contributes to decreased ADAMTS-13 activity in Koreans. The frequency of the C1423T allele of the ADAMTS13 gene was studied along with measuring plasma ADAMTS-13 activity in 250 healthy Korean individuals. The allele frequency of C1423T polymorphism was 4%, and the median activity of CT type was 107 (69-143)%, which was lower than in controls with the CC genotype [118 (48-197)%, (p=0.021)]. Therefore, the Pro475Ser polymorphism seems to be popular in the Korean population, and attenuates ADAMTS-13 plasma activity.