Merck
  • Novel tyrosinase inhibitors from liquid culture of Neolentinus lepideus.
Novel tyrosinase inhibitors from liquid culture of Neolentinus lepideus.
Bioscience, biotechnology, and biochemistry (2018-01-04)
Atsushi Ishihara, Yuri Ide, Tomohiro Bito, Naoki Ube, Naoki Endo, Kozue Sotome, Nitaro Maekawa, Kotomi Ueno, Akira Nakagiri
ABSTRACT
Tyrosinase is the key enzyme that controls melanin formation in the human skin. We performed a screening of 96 extracts of mushroom cultures and fruiting bodies for examining their inhibitory activity against mushroom tyrosinase. The ethyl acetate extracts of culture filtrate of Neolentinus lepideus exhibited the strongest inhibitory activity. The active compounds 1 and 2 were purified by repeated chromatographic separations from the extract. On the basis of spectroscopic analyses, 1 and 2 were identified to be 1,3-dihydroisobenzofuran-4,5,7-triol and 5-methoxy-1,3-dihydroisobenzofuran-4,7-diol, respectively. Lineweaver-Burk plot of the enzyme reaction in the presence of 1 indicated that 1 was a potent competitive inhibitor. The respective IC