Merck
  • Autophagy induces apoptosis and death of T lymphocytes in the spleen of pigs infected with CSFV.
Autophagy induces apoptosis and death of T lymphocytes in the spleen of pigs infected with CSFV.
Scientific reports (2017-10-21)
Hongchao Gou, Mingqiu Zhao, Shuangqi Fan, Jin Yuan, Jiedan Liao, Wencheng He, Hailuan Xu, Jinding Chen
ABSTRACT
Lymphocyte depletion and immunosuppression are typical clinical characteristics of pigs infected with classical swine fever virus (CSFV). The apoptosis of virus-infected and bystander cells plays a role in the immunopathology of classical swine fever (CSF). Here, we offer the first evidence that autophagy is involved in apoptosis and death of T lymphocytes in the spleen of pigs infected with CSFV. Using immunohistochemical assays, we observed that more LC3II-positive cells appear in the T-cell zone of spleens. Spleen cell apoptosis was demonstrated using flow cytometry and TUNEL staining. Confocal immunofluorescence revealed that partial LC3II-positive cells were simultaneously TUNEL-positive. By cultivating spleen cells ex vivo, we demonstrated that the inhibition of autophagy by 3-MA treatment inhibited apoptosis and death of T lymphocytes caused by CSFV infection but did not have this effect  on B lymphocytes. Further observations demonstrated that uninfected cells in the spleen were also undergoing autophagy in vivo. In summary, these results linked autophagy with the apoptosis and cell death of splenic T cells, providing a new outlook to understand the mechanism of T lymphocyte depletion and immunosuppression during CSF.
Sigma-Aldrich
Anti-SQSTM1 antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.