Merck
  • The mevalonate pathway regulates primitive streak formation via protein farnesylation.
The mevalonate pathway regulates primitive streak formation via protein farnesylation.
Scientific reports (2016-11-25)
Yoshimi Okamoto-Uchida, Ruoxing Yu, Norio Miyamura, Norie Arima, Mari Ishigami-Yuasa, Hiroyuki Kagechika, Suguru Yoshida, Takamitsu Hosoya, Makiko Nawa, Takeshi Kasama, Yoichi Asaoka, Reiner Wimmer Alois, Ulrich Elling, Josef M Penninger, Sachiko Nishina, Noriyuki Azuma, Hiroshi Nishina
ABSTRACT
The primitive streak in peri-implantation embryos forms the mesoderm and endoderm and controls cell differentiation. The metabolic cues regulating primitive streak formation remain largely unknown. Here we utilised a mouse embryonic stem (ES) cell differentiation system and a library of well-characterised drugs to identify these metabolic factors. We found that statins, which inhibit the mevalonate metabolic pathway, suppressed primitive streak formation in vitro and in vivo. Using metabolomics and pharmacologic approaches we identified the downstream signalling pathway of mevalonate and revealed that primitive streak formation requires protein farnesylation but not cholesterol synthesis. A tagging-via-substrate approach revealed that nuclear lamin B1 and small G proteins were farnesylated in embryoid bodies and important for primitive streak gene expression. In conclusion, protein farnesylation driven by the mevalonate pathway is a metabolic cue essential for primitive streak formation.
Sigma-Aldrich
Monoclonal Anti-Neurofilament 68 antibody produced in mouse, clone NR4, ascites fluid
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, ChemiconĀ®, from mouse