Merck
  • Armc5 deletion causes developmental defects and compromises T-cell immune responses.
Armc5 deletion causes developmental defects and compromises T-cell immune responses.
Nature communications (2017-02-09)
Yan Hu, Linjiang Lao, Jianning Mao, Wei Jin, Hongyu Luo, Tania Charpentier, Shijie Qi, Junzheng Peng, Bing Hu, Mieczyslaw Martin Marcinkiewicz, Alain Lamarre, Jiangping Wu
ABSTRACT
Armadillo repeat containing 5 (ARMC5) is a cytosolic protein with no enzymatic activities. Little is known about its function and mechanisms of action, except that gene mutations are associated with risks of primary macronodular adrenal gland hyperplasia. Here we map Armc5 expression by in situ hybridization, and generate Armc5 knockout mice, which are small in body size. Armc5 knockout mice have compromised T-cell proliferation and differentiation into Th1 and Th17 cells, increased T-cell apoptosis, reduced severity of experimental autoimmune encephalitis, and defective immune responses to lymphocytic choriomeningitis virus infection. These mice also develop adrenal gland hyperplasia in old age. Yeast 2-hybrid assays identify 16 ARMC5-binding partners. Together these data indicate that ARMC5 is crucial in fetal development, T-cell function and adrenal gland growth homeostasis, and that the functions of ARMC5 probably depend on interaction with multiple signalling pathways.
Product Description
Roche
Anti-HA-Biotin, High Affinity (3F10), from rat IgG1
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Pricing and availability is not currently available.