Merck
  • Erratum to "Resveratrol Attenuates Copper-Induced Senescence by Improving Cellular Proteostasis".
Erratum to "Resveratrol Attenuates Copper-Induced Senescence by Improving Cellular Proteostasis".
Oxidative medicine and cellular longevity (2017-11-18)
Liliana Matos, Alexandra Monteiro Gouveia, Henrique Almeida
ABSTRACT
[This corrects the article DOI: 10.1155/2017/3793817.].
Sigma-Aldrich
Anti-DNP antibody produced in rabbit, whole antiserum