Merck
  • Transmembrane and Coiled-Coil Domain 1 Impairs the AKT Signaling Pathway in Urinary Bladder Urothelial Carcinoma: A Characterization of a Tumor Suppressor.
Transmembrane and Coiled-Coil Domain 1 Impairs the AKT Signaling Pathway in Urinary Bladder Urothelial Carcinoma: A Characterization of a Tumor Suppressor.
Clinical cancer research : an official journal of the American Association for Cancer Research (2017-10-04)
Chien-Feng Li, Wen-Ren Wu, Ti-Chun Chan, Yu-Hui Wang, Lih-Ren Chen, Wen-Jeng Wu, Bi-Wen Yeh, Shih-Shin Liang, Yow-Ling Shiue
ABSTRACT
Purpose: Urinary bladder urothelial carcinoma (UBUC) is a common malignant disease in developed countries. Cell-cycle dysregulation resulting in uncontrolled cell proliferation has been associated with UBUC development. This study aimed to explore the roles of TMCO1 in UBUCs.Experimental Design: Data mining, branched DNA assay, immunohistochemistry, xenograft, cell culture, quantitative RT-PCR, immunoblotting, stable and transient transfection, lentivirus production and stable knockdown, cell-cycle, cell viability and proliferation, soft-agar, wound-healing, transwell migration and invasion, coimmunoprecipitation, immunocytochemistry, and AKT serine/threonine kinase (AKT) activity assays and site-directed mutagenesis were used to study TMCO1 involvement in vivo and in vitroResults: Data mining identified that the TMCO1 transcript was downregulated during the progression of UBUCs. In distinct UBUC-derived cell lines, changes in TMCO1 levels altered the cell-cycle distribution, cell viability, cell proliferation, and colony formation and modulated the AKT pathway. TMCO1 recruited the PH domain and leucine-rich repeat protein phosphatase 2 (PHLPP2) to dephosphorylate pAKT1(serine 473) (S473). Mutagenesis at S60 of the TMCO1 protein released TMCO1-induced cell-cycle arrest and restored the AKT pathway in BFTC905 cells. Stable TMCO1 (wild-type) overexpression suppressed, whereas T33A and S60A mutants recovered, tumor size in xenograft mice.Conclusions: Clinical associations, xenograft mice, and in vitro indications provide solid evidence that the TMCO1 gene is a novel tumor suppressor in UBUCs. TMCO1 dysregulates cell-cycle progression via suppression of the AKT pathway, and S60 of the TMCO1 protein is crucial for its tumor-suppressor roles. Clin Cancer Res; 23(24); 7650-63. ©2017 AACR.
Sigma-Aldrich
QCM ECMatrix Cell Invasion Assay, 24-well (8 µm), fluorimetric, The Cell Invasion Assay Kit kuses a 24 well plate with 8 um pores, which is ideal for evaluation of invasive tumor cells.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O