Merck
  • Biomarker monitoring of controlled dietary acrylamide exposure indicates consistent human endogenous background.
Biomarker monitoring of controlled dietary acrylamide exposure indicates consistent human endogenous background.
Archives of toxicology (2017-05-24)
Katharina Goempel, Laura Tedsen, Meike Ruenz, Tamara Bakuradze, Dorothea Schipp, Jens Galan, Gerhard Eisenbrand, Elke Richling
ABSTRACT
The aim of the present study was to explore the relation of controlled dietary acrylamide (AA) intake with the excretion of AA-related urinary mercapturic acids (MA), N-acetyl-S-(carbamoylethyl)-L-cysteine (AAMA) and N-acetyl-S-(1-carbamoyl-2-hydroxyethyl)-L-cysteine (GAMA). Excretion kinetics of these short-term exposure biomarkers were monitored under strictly controlled conditions within a duplicate diet human intervention study. One study arm (group A, n = 6) ingested AA via coffee (0.15-0.17 µg/kg bw) on day 6 and in a meal containing an upper exposure level of AA (14.1-15.9 μg/kg bw) on day 10. The other arm (group B) was on AA minimized diet (washout, 0.05-0.06 µg/kg bw) throughout the whole 13-day study period. On day 6, these volunteers ingested
Sigma-Aldrich
Pentafluorophenyl isothiocyanate, ≥97.0% (GC)