- Gradients in Wall Mechanics and Polysaccharides along Growing Inflorescence Stems.
Gradients in Wall Mechanics and Polysaccharides along Growing Inflorescence Stems.
Plant physiology (2017-11-01)
Pyae Phyo, Tuo Wang, Sarah N Kiemle, Hugh O'Neill, Sai Venkatesh Pingali, Mei Hong, Daniel J Cosgrove
PMID29084904
ABSTRACT
At early stages of Arabidopsis (
Sigma-Aldrich
Murashige and Skoog Basal Salt Mixture (MS), powder, suitable for plant cell culture