Merck
Exploring the effect of fluorinated anions on the CO
Physical chemistry chemical physics : PCCP (2017-10-24)
Andreia S L Gouveia, Liliana C Tomé, Elena I Lozinskaya, Alexander S Shaplov, Yakov S Vygodskii, Isabel M Marrucho
ABSTRACT
The CO
Sigma-Aldrich
Bromoethane, reagent grade, 98%