Merck
  • Sulphur dioxide suppresses inflammatory response by sulphenylating NF-κB p65 at Cys
Sulphur dioxide suppresses inflammatory response by sulphenylating NF-κB p65 at Cys
Clinical science (London, England : 1979) (2017-09-25)
Siyao Chen, Yaqian Huang, Zhiwei Liu, Wen Yu, Heng Zhang, Kun Li, Xiaoqi Yu, Chaoshu Tang, Bin Zhao, Junbao Du, Hongfang Jin
ABSTRACT
The present study was designed to investigate whether endogenous sulphur dioxide (SO
Sigma-Aldrich
Bay 11-7082, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%
Sigma-Aldrich
MISSION® esiRNA, targeting human GPT