Merck
  • Thrombin-activatable fibrinolysis inhibitor in human abdominal aortic aneurysm disease.
Thrombin-activatable fibrinolysis inhibitor in human abdominal aortic aneurysm disease.
Journal of thrombosis and haemostasis : JTH (2017-08-24)
K I Bridge, L Bollen, J Zhong, M Hesketh, F L Macrae, A Johnson, H Philippou, D J Scott, A Gils, R A S Ariёns
ABSTRACT
Essentials Patients with abdominal aortic aneurysms (AAA) develop dense clots that are resistant to lysis. This study explores the role of thrombin-activatable fibrinolysis inhibitor (TAFI) in human AAA. There is evidence of chronically increased TAFI activation in patients with AAA. TAFI may represent a pharmacological target for cardiovascular risk reduction in AAA. Background Intra-luminal thrombosis is a key factor in growth of abdominal aortic aneurysms (AAAs). Patients with AAA form dense clots that are resistant to fibrinolysis. Thrombin-activatable fibrinolysis inhibitor (TAFI) has been shown to influence AAA development in murine models. Objective The aim of this study is to characterize the role of TAFI in human AAA. Methods Plasma levels of TAFI, TAFI activation peptide (TAFI-AP), activated/inactivated TAFI (TAFIa/ai) and plasmin-α2-antiplasmin complex were measured by ELISAs in patients with AAA (n = 202) and controls (n = 188). Results TAFIa/ai and TAFI-AP levels were higher in patients than controls (median [IQR], 20.3 [14.6-32.8] ng mL