Merck
  • Antihistaminic and anticholinergic activities of mequitazine in comparison with clemizole.
Antihistaminic and anticholinergic activities of mequitazine in comparison with clemizole.
The Journal of pharmacy and pharmacology (1988-09-01)
I Martinez-Mir, L Estañ, E Rubio, F J Morales-Olivas
ABSTRACT
The antihistamine and anticholinergic properties of mequitazine have been investigated and compared with those of clemizole. Both mequitazine and clemizole antagonized the effect of histamine in guinea-pig ileum competitively, the pA2 values calculated by Schild plot were 9.95 +/- 0.44 for mequitazine and 10.54 +/- 0.44 for clemizole. Mequitazine at 10(-7) M produced a parallel shift of the dose-response curve to acetylcholine in the rat duodenum, clemizole and the lower doses of mequitazine failed to modify the effect of acetylcholine. The potency of mequitazine and clemizole as H1-histamine blockers is similar, but only mequitazine at highest concentration used showed anticholinergic activity.
Sigma-Aldrich
Mequitazine, ≥98% (HPLC)