Merck
Inhibition of Mitochondrial Bioenergetics by Esterase-Triggered COS/H
ACS chemical biology (2017-06-15)
Andrea K Steiger, Michela Marcatti, Csaba Szabo, Bartosz Szczesny, Michael D Pluth
ABSTRACT
Hydrogen sulfide (H