Merck
  • The Golgi matrix protein giantin is required for normal cilia function in zebrafish.
The Golgi matrix protein giantin is required for normal cilia function in zebrafish.
Biology open (2017-05-27)
Dylan J M Bergen, Nicola L Stevenson, Roderick E H Skinner, David J Stephens, Christina L Hammond
ABSTRACT
The Golgi is essential for glycosylation of newly synthesised proteins including almost all cell-surface and extracellular matrix proteoglycans. Giantin, encoded by the
Sigma-Aldrich
MISSION® esiRNA, targeting human GOLGB1
Sigma-Aldrich
SANT-1, ≥98% (HPLC), powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid