Merck
  • Functional annotation of a novel NFKB1 promoter polymorphism that increases risk for ulcerative colitis.
Functional annotation of a novel NFKB1 promoter polymorphism that increases risk for ulcerative colitis.
Human molecular genetics (2003-11-14)
Amir S Karban, Toshihiko Okazaki, Carolien I M Panhuysen, Thomas Gallegos, James J Potter, Joan E Bailey-Wilson, Mark S Silverberg, Richard H Duerr, Judy H Cho, Peter K Gregersen, Yuqiong Wu, Jean-Paul Achkar, Themistocles Dassopoulos, Esteban Mezey, Theodore M Bayless, Franklin J Nouvet, Steven R Brant
ABSTRACT
Nuclear Factor-kappaB (NF-kappaB) is a major transcription regulator of immune response, apoptosis and cell-growth control genes, and is upregulated in inflammatory bowel disease (IBD), both ulcerative colitis (UC) and Crohn's disease. The NFKB1 gene encodes the NF-kappaB p105/p50 isoforms. Genome-wide screens in IBD families show evidence for linkage on chromosome 4q where NFKB1 maps. We sequenced the NFKB1 promoter, exon 1 and all coding exons in 10 IBD probands and two controls, and identified six nucleotide variants, including a common insertion/deletion promoter polymorphism (-94ins/delATTG). Using pedigree-based transmission disequilibrium tests, we observed modest evidence for linkage disequilibrium (LD), independent of linkage, between the -94delATTG allele and UC in 131 out of 235 IBD pedigrees with UC offspring (P=0.047-0.052). This allele was also more frequent in the 156 non-Jewish UC probands from the 235 IBD pedigrees than in 149 non-Jewish controls (P=0.015). The -94delATTG association with UC was replicated in a second set of 258 unrelated, non-Jewish UC cases and 653 new, non-Jewish controls (P=0.021). Nuclear proteins from normal human colon tissue and colonic cell lines, but not ileal tissue, showed significant binding to -94insATTG but not to -94delATTG containing oligonucleotides. NFKB1 promoter/exon 1 luciferase reporter plasmid constructs containing the -94delATTG allele and transfected into either HeLa or HT-29 cell lines showed less promoter activity than comparable constructs containing the -94insATTG allele. Therefore, we have identified the first potentially functional polymorphism of NFKB1 and demonstrated its genetic association with a common human disease, ulcerative colitis.