Merck
  • Sustained release docetaxel-incorporated lipid nanoparticles with improved pharmacokinetics for oral and parenteral administration.
Sustained release docetaxel-incorporated lipid nanoparticles with improved pharmacokinetics for oral and parenteral administration.
Journal of microencapsulation (2017-05-31)
Omer Salman Qureshi, Hyung-Seo Kim, Alam Zeb, Jin-Seok Choi, Hoo-Seong Kim, Jung-Eun Kwon, Myung-Sic Kim, Jong-Ho Kang, Chongsuk Ryou, Jeong-Sook Park, Jin-Ki Kim
ABSTRACT
The aim of this study was to develop docetaxel-incorporated lipid nanoparticles (DTX-NPs) to improve the pharmacokinetic behaviour of docetaxel (DTX) after oral and parenteral administration via sustained release. DTX-NPs were prepared by nanotemplate engineering technique with palmityl alcohol as a solid lipid and Tween-40/Span-40/Myrj S40 as a surfactants mixture. Spherical DTX-NPs below 100 nm were successfully prepared with a narrow particle size distribution, 96% of incorporation efficiency and 686 times increase in DTX solubility. DTX-NPs showed a sustained release over 24 h in phosphate-buffered saline and simulated gastric and intestinal fluids, while DTX-micelles released DTX completely within 12 h. The half-maximal inhibitory concentration (IC
Sigma-Aldrich
Sorbitan monopalmitate