Merck
  • MOB1-YAP1/TAZ-NKX2.1 axis controls bronchioalveolar cell differentiation, adhesion and tumour formation.
MOB1-YAP1/TAZ-NKX2.1 axis controls bronchioalveolar cell differentiation, adhesion and tumour formation.
Oncogene (2017-03-28)
K Otsubo, H Goto, M Nishio, K Kawamura, S Yanagi, W Nishie, T Sasaki, T Maehama, H Nishina, K Mimori, T Nakano, H Shimizu, T W Mak, K Nakao, Y Nakanishi, A Suzuki
ABSTRACT
Mps One Binder Kinase Activator (MOB)1A/1B are core components of the Hippo pathway. These proteins, which coactivate LArge Tumour Suppressor homologue kinases, are also tumour suppressors. To investigate MOB1A/B's roles in normal physiology and lung cancer, we generated doxycycline (Dox)-inducible, bronchioalveolar epithelium-specific, null mutations of MOB1A/B in mice (SPC-rtTA/(tetO)
Sigma-Aldrich
3-(Biphenyl-4-yl)-5-(4-tert-butylphenyl)-4-phenyl-4H-1,2,4-triazole, 97%