Merck
  • Functional and molecular characterization of voltage gated sodium channel Na
Functional and molecular characterization of voltage gated sodium channel Na
Theriogenology (2017-02-09)
Dharmendra Singh Chauhan, Dilip Kumar Swain, Nadeem Shah, Hanuman Prasad Yadav, Udayraj P Nakade, Vijay Kumar Singh, Rajesh Nigam, Sarvajeet Yadav, Satish Kumar Garg
ABSTRACT
The aim of our study was to characterize the voltage gated sodium channel Na
Sigma-Aldrich
Veratridine, ≥90% (HPLC), powder