Merck
Cytotoxic and Enzyme Inhibitory Potential of Two
Frontiers in pharmacology (2017-06-08)
Sengul Uysal, Gokhan Zengin, Marcello Locatelli, Mir B Bahadori, Andrei Mocan, Giuseppe Bellagamba, Elisa De Luca, Adriano Mollica, Abdurrahman Aktumsek
ABSTRACT
In this work, the biological and chemical fingerprints of three extracts (ethyl acetate, methanol, and water) from two