Merck
  • Macrophage migration inhibitory factor interacts with thioredoxin-interacting protein and induces NF-κB activity.
Macrophage migration inhibitory factor interacts with thioredoxin-interacting protein and induces NF-κB activity.
Cellular signalling (2017-03-23)
Mi Jeong Kim, Won Sam Kim, Dong Oh Kim, Jae-Eun Byun, Hangsak Huy, Soo Yun Lee, Hae Young Song, Young-Jun Park, Tae-Don Kim, Suk Ran Yoon, Eun-Ji Choi, Hyunjung Ha, Haiyoung Jung, Inpyo Choi
ABSTRACT
The nuclear factor kappa B (NF-κB) pathway is pivotal in controlling survival and apoptosis of cancer cells. Macrophage migration inhibitory factor (MIF), a cytokine that regulates the immune response and tumorigenesis under inflammatory conditions, is upregulated in various tumors. However, the intracellular functions of MIF are unclear. In this study, we found that MIF directly interacted with thioredoxin-interacting protein (TXNIP), a tumor suppressor and known inhibitor of NF-κB activity, and MIF significantly induced NF-κB activation. MIF competed with TXNIP for NF-κB activation, and the intracellular MIF induced NF-κB target genes, including c-IAP2, Bcl-xL, ICAM-1, MMP2 and uPA, by inhibiting the interactions between TXNIP and HDACs or p65. Furthermore, we identified the interaction motifs between MIF and TXNIP via site-directed mutagenesis of their cysteine (Cys) residues. Cys
Sigma-Aldrich
MISSION® esiRNA, targeting human MIF
Sigma-Aldrich
MISSION® esiRNA, targeting human TXNIP