Merck
  • Response Patterns of GABAergic Neurons in the Anterior Piriform Cortex of Awake Mice.
Response Patterns of GABAergic Neurons in the Anterior Piriform Cortex of Awake Mice.
Cerebral cortex (New York, N.Y. : 1991) (2016-06-03)
Rongfeng Hu, Juen Zhang, Minmin Luo, Ji Hu
ABSTRACT
Local inhibition by γ-amino butyric acid (GABA)-containing neurons is of vital importance for the operation of sensory cortices. However, the physiological response patterns of cortical GABAergic neurons are poorly understood, especially in the awake condition. Here, we utilized the recently developed optical tagging technique to specifically record GABAergic neurons in the anterior piriform cortex (aPC) in awake mice. The identified aPC GABAergic neurons were stimulated with robotic delivery of 32 distinct odorants, which covered a broad range of functional groups. We found that aPC GABAergic neurons could be divided into 4 types based on their response patterns. Type I, type II, and type III neurons displayed broad excitatory responses to test odorants with different dynamics. Type I neurons were constantly activated during odorant stimulation, whereas type II neurons were only transiently activated at the onset of odorant delivery. In addition, type III neurons displayed transient excitatory responses both at the onset and termination of odorant presentation. Interestingly, type IV neurons were broadly inhibited by most of the odorants. Taken together, aPC GABAergic neurons adopt different strategies to affect the cortical circuitry. Our results will allow for better understanding of the role of cortical GABAergic interneurons in sensory information processing.