Merck
  • Furosemide depresses the presynaptic fiber volley and modifies frequency-dependent axonal excitability in rat hippocampus.
Furosemide depresses the presynaptic fiber volley and modifies frequency-dependent axonal excitability in rat hippocampus.
Journal of neurophysiology (2017-01-20)
Mogens Andreasen, Steen Nedergaard
ABSTRACT
The loop diuretic furosemide is known to have anticonvulsant effects, believed to be exerted through blockade of glial Na
Sigma-Aldrich
VU0240551, ≥98% (HPLC)