- Postpolymerization modification of a hydroxy monolith precursor. Part I. Epoxy alkane and octadecyl isocyanate modified poly (hydroxyethyl methacrylate-co-pentaerythritol triacrylate) monolithic capillary columns for reversed-phase capillary electrochromatography.
Postpolymerization modification of a hydroxy monolith precursor. Part I. Epoxy alkane and octadecyl isocyanate modified poly (hydroxyethyl methacrylate-co-pentaerythritol triacrylate) monolithic capillary columns for reversed-phase capillary electrochromatography.
Electrophoresis (2016-09-10)
Shantipriya Khadka, Ziad El Rassi
PMID27611883
ABSTRACT
A novel precursor monolithic capillary column referred to as "hydroxy monolith" or OHM was prepared by the in situ copolymerization of hydroxyethylmethacrylate (HEMA) with pentaerythritol triacrylate (PETA) yielding the neutral poly(HEMA-co-PETA) monolith. The neutral precursor OHM capillary thus obtained was subjected to postpolymerization modifications of the hydroxyl functional groups present on its surface with 1,2-epoxyalkanes catalyzed by boron trifluoride (BF
Sigma-Aldrich
Pentaerythritol tetraacrylate, Pentaerythritol triacrylate, and Trimethylolpropane triacrylate mixture