Merck
  • Dysregulation of c-Myb Pathway by Aberrant Expression of Proto-oncogene MYB Provides the Basis for Malignancy in Adult T-cell Leukemia/lymphoma Cells.
Dysregulation of c-Myb Pathway by Aberrant Expression of Proto-oncogene MYB Provides the Basis for Malignancy in Adult T-cell Leukemia/lymphoma Cells.
Clinical cancer research : an official journal of the American Association for Cancer Research (2016-11-04)
Kazumi Nakano, Kaoru Uchimaru, Atae Utsunomiya, Kazunari Yamaguchi, Toshiki Watanabe
ABSTRACT
Adult T-cell leukemia/lymphoma (ATLL) is an aggressive human T-cell malignancy induced by human T-lymphotrophic virus-1 (HTLV-1) infection. The genetic alterations in infected cells that lead to transformation have not been completely elucidated, thus hindering the identification of effective therapeutic targets for ATL. Here, we present the first assessment of MYB proto-oncogene dysregulation in ATL and an exploration of its role in the onset of ATL. We investigated the expression patterns of MYB splicing variants in ATL. The molecular characteristics of the c-Myb-9A isoform, which was overexpressed in ATL cells, were examined using chromatin immunoprecipitation and promoter assays. We further examined the biologic impacts of abnormal c-Myb overexpression in ATL using overall c-Myb knockdown with shRNA or c-Myb-9A knockdown with morpholino oligomers. Both total c-Myb and c-Myb-9A, which exhibited strong transforming activity, were overexpressed in ATL cells in a leukemogenesis- and progression-dependent manner. Knockdown of either total c-Myb or c-Myb-9A induced ATL cell death. c-Myb transactivates nine genes that encode essential regulators of cell proliferation and NF-κB signaling. c-Myb-9A induced significantly stronger transactivation of all tested genes and stronger NF-κB activation compared with wild-type c-Myb. Our data demonstrate that c-Myb pathway overactivation caused by unbalanced c-Myb-9A overexpression is associated with disorders in cellular homeostasis and consequently, accelerated transformation, cell proliferation, and malignancy in ATL cells. These data support the notion of the c-Myb pathway as a promising new therapeutic target for ATL. Clin Cancer Res; 22(23); 5915-28. ©2016 AACR.
Sigma-Aldrich
IgG from mouse serum, reagent grade, ≥95% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Anti-NFκB Antibody, p65 subunit, active subunit, clone 12H11, clone 12H11, Chemicon®, from mouse
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)