Merck
Peripherin-IgG association with neurologic and endocrine autoimmunity.
Journal of autoimmunity (2010-01-12)
Jayne L Chamberlain, Sean J Pittock, Anna-Maria Oprescu, Carissa Dege, Metha Apiwattanakul, Thomas J Kryzer, Vanda A Lennon
ABSTRACT
Peripherin-IgG has been reported a pertinent autoantibody in non-obese type 1 diabetic (NOD) mice. However, it has not previously been recognized in any human disease. In blinded evaluation of serum for markers of neurological autoimmunity in a high-volume diagnostic laboratory, we incidentally identified 26 patients (61% female) with an IgG that bound selectively to neural elements in enteric ganglia, sympathetic nerve trunks and discrete nerve tracts in mid-brain and hind-brain. The target antigen was identified as peripherin, a 55kDa - type III intermediate filament protein. Review of clinical histories revealed that 54% of seropositive patients had dysautonomia (predominantly gastrointestinal dysmotility), 30% had neuropathies with varied sensory symptoms and 35% had clinical or serological evidence of endocrinopathy (type 1 diabetes, thyroiditis or premature ovarian failure). Collectively, 73% had autonomic dysfunction or endocrinopathy. None of 173 healthy subjects was seropositive. Subsequent western blot evaluation of archival sera from patients with small fiber/autonomic neuropathies (with or without endocrinopathy) revealed a 33% seropositivity rate for peripherin-IgG. Our further demonstration that peripherin-immunoreactive autonomic fibers in pancreas, thyroid and ovary are juxtaposed to endocrine epithelium, complement our clinical observations in suggesting that neuronal elements may be a pertinent initial target for immune attack in multiple forms of endocrine autoimmunity (intermolecular epitope spreading). It remains to be determined whether or not peripherin-IgG is predictive for development of small fiber neuropathy (autonomic or somatic).
Sigma-Aldrich
Anti-Peripherin Antibody, clone 8G2, culture supernatant, clone 8G2, Chemicon®
Millipore
ProteoExtract® Subcellular Proteome Extraction Kit
Sigma-Aldrich
Anti-Neurofilament M (145 kDa) Antibody, CT, serum, Chemicon®
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP)−Cy3 antibody, Mouse monoclonal, clone G-A-5, purified from hybridoma cell culture