Merck
  • Identification of In-Chain-Functionalized Compounds and Methyl-Branched Alkanes in Cuticular Waxes of Triticum aestivum cv. Bethlehem.
Identification of In-Chain-Functionalized Compounds and Methyl-Branched Alkanes in Cuticular Waxes of Triticum aestivum cv. Bethlehem.
PloS one (2016-11-08)
Radu C Racovita, Reinhard Jetter
ABSTRACT
In this work, cuticular waxes from flag leaf blades and peduncles of Triticum aestivum cv. Bethlehem were investigated in search for novel wax compounds. Seven wax compound classes were detected that had previously not been reported, and their structures were elucidated using gas chromatography-mass spectrometry of various derivatives. Six of the classes were identified as series of homologs differing by two methylene units, while the seventh was a homologous series with homologs with single methylene unit differences. In the waxes of flag leaf blades, secondary alcohols (predominantly C27 and C33), primary/secondary diols (predominantly C28) and esters of primary/secondary diols (predominantly C50, combining C28 diol with C22 acid) were found, all sharing similar secondary hydroxyl group positions at and around C-12 or ω-12. 7- and 8-hydroxy-2-alkanol esters (predominantly C35), 7- and 8-oxo-2-alkanol esters (predominantly C35), and 4-alkylbutan-4-olides (predominantly C28) were found both in flag leaf and peduncle wax mixtures. Finally, a series of even- and odd-numbered alkane homologs was identified in both leaf and peduncle waxes, with an internal methyl branch preferentially on C-11 and C-13 of homologs with even total carbon number and on C-12 of odd-numbered homologs. Biosynthetic pathways are suggested for all compounds, based on common structural features and matching chain length profiles with other wheat wax compound classes.
Sigma-Aldrich
Tetracosane, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mg
1 Available to ship on 07 January 2025
Details...
MXP 2,412.00
500 mg
1 Available to ship on 07 January 2025
Details...
MXP 8,880.00
1 g
1 Available to ship on 07 January 2025
Details...
MXP 12,068.00
5 g
1 Estimated to ship on 09 March 2025
MXP 53,406.00