Merck
  • Synthesis and biological evaluation of N4-(hetero)arylsulfonylquinoxalinones as HIV-1 reverse transcriptase inhibitors.
Synthesis and biological evaluation of N4-(hetero)arylsulfonylquinoxalinones as HIV-1 reverse transcriptase inhibitors.
Bioorganic & medicinal chemistry (2009-03-10)
Bailing Xu, Yan Sun, Ying Guo, Yingli Cao, Tao Yu
ABSTRACT
A series of novel N(4)-(hetero)arylsulfonylquinoxalinone derivatives were prepared in a straight and efficient way. Of all the synthesized compounds, five compounds exhibited potent anti-HIV-1 replication activities with IC(50) value at the level of 10(-7) mol/L. Preliminary structure-activity relationships were studied in details and that will shed light on the discovery of more potent non-nucleoside reverse-transcriptase inhibitors.