Merck
  • Interleukin-8 is a prognostic indicator in human hilar cholangiocarcinoma.
Interleukin-8 is a prognostic indicator in human hilar cholangiocarcinoma.
International journal of clinical and experimental pathology (2015-09-05)
Qi Sun, Fanni Li, Fengkai Sun, Jun Niu
ABSTRACT
Interleukin-8 (IL-8), matrix metalloproteinase-9 (MMP-9) and neovascularization have been implicated to be associated with biological processes, especially cancer progression. However, few studies have investigated the role of IL-8 in human hilar cholangiocarcinoma. In this study we detected the expression of IL-8 combined with MMP-9 and microvessel density (MVD) in hilar cholangiocarcinoma to evaluate their clinicopathological significance and prognostic value. A total of 62 patients with hilar cholangiocarcinoma who underwent curative surgery were enrolled in this study. The expression of IL-8, MMP-9 and MVD were examined immunohistochemically. The correlation of IL-8 with MMP-9 expression, MVD, clinicopathological features and survival time of patients were then analyzed. Expression of IL-8 was observed in 56.5% tumors, which was related to advanced TNM stage (P = 0.026) and tumor recurrence (P = 0.018). IL-8 had a positive correlation with MMP-9 expression and MVD. Furthermore, patients with high IL-8 expression had a significantly shorter overall survival than those with low IL-8 expression (P = 0.01). Multivariate analysis confirmed IL-8 as an independent prognostic factor (P = 0.005). In conclusion, IL-8 expression significantly correlated with MMP-9 expression and MVD, and IL-8 was a valuable prognostic factor for human hilar cholangiocarcinoma.
Sigma-Aldrich
IL-8 (77aa) (CXCL8) human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
IL-8 (72AA) human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)