Merck
  • Efficient Semitransparent Perovskite Solar Cells with Graphene Electrodes.
Efficient Semitransparent Perovskite Solar Cells with Graphene Electrodes.
Advanced materials (Deerfield Beach, Fla.) (2015-05-15)
Peng You, Zhike Liu, Qidong Tai, Shenghua Liu, Feng Yan
ABSTRACT
Semitransparent perovskite solar cells are prepared by laminating graphene transparent electrodes on the top for the first time. The device performance is optimized by improving the conductivity of the graphene electrodes and the contact between the graphene and the perovskite active layers during the lamination process. The devices show high power conversion efficiencies when they are illuminated from both sides.
Sigma-Aldrich
Graphene/PEDOT:PSS hybrid ink, dispersion in DMF, avg. no. of layers, 1 ‑ 3