Merck
Microencapsulation of osmium tetroxide in polyurea.
Organic letters (2003-01-17)
Steven V Ley, Chandrashekar Ramarao, Ai-Lan Lee, Niels Østergaard, Stephen C Smith, Ian M Shirley
ABSTRACT
[reaction: see text] Osmium tetroxide has been microencapsulated in a polyurea matrix using an in situ interfacial polymerization approach. These microcapsules have been effectively used as recoverable and reusable catalysts in the dihydroxylation of olefins
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading